Invitrogen
Manufacturer Code:PA587454
Catalog # PIPA587454
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 1-300 of human PKN1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: PAK-1; Protease-activated kinase 1; Protein kinase C-like 1; Protein kinase C-like PKN; protein kinase C-related kinase 1; protein kinase PKN; Protein kinase PKN-alpha; Protein-kinase C-related kinase 1; serine-threonine kinase N; Serine-threonine protein kinase N; serine/threonine kinase 3; serine/threonine protein kinase N; Serine/threonine-protein kinase N1
Gene Aliases: DBK; F730027O18Rik; PAK-1; PAK1; PKN; PKN-ALPHA; PKN1; PRK1; PRKCL1; Stk3
UniProt ID: (Human) Q16512, (Rat) Q63433, (Mouse) P70268
Entrez Gene ID: (Human) 5585, (Rat) 29355, (Mouse) 320795
Molecular Function: annexin calcium-binding protein calmodulin intracellular calcium-sensing protein kinase non-receptor serine/threonine protein kinase protein kinase transfer/carrier protein transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.