Novus Biologicals
Manufacturer Code:NBP298761AF750
Catalog # NB105619
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human PKN1. |
Conjugate | Alexa Fluor 750 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DBK EC 2.7.11 MGC46204 PAK1Serine-threonine protein kinase N PKNPKN-ALPHA PRK1PAK-1 PRKCL1protein kinase C-related kinase 1 Protease-activated kinase 1 Protein kinase C-like 1EC 2.7.11.13 Protein kinase C-like PKN protein kinase N1 Protein kinase PKN-alpha Protein-kinase C-related kinase 1 serine/threonine protein kinase N serine/threonine-protein kinase N1 serine-threonine kinase N; PAK-1; Protease-activated kinase 1; Protein kinase C-like 1; Protein kinase C-like PKN; protein kinase C-related kinase 1; Protein kinase PKN-alpha; Protein-kinase C-related kinase 1; serine-threonine kinase N; Serine-threonine protein kinase N; serine/threonine protein kinase N; Serine/threonine-protein kinase N1
Gene Aliases: DBK; PAK-1; PAK1; PKN; PKN-ALPHA; PKN1; PRK1; PRKCL1
UniProt ID: (Human) Q16512
Entrez Gene ID: (Human) 5585
Molecular Function: annexin calcium-binding protein calmodulin intracellular calcium-sensing protein kinase non-receptor serine/threonine protein kinase protein kinase transfer/carrier protein transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.