Novus Biologicals
Manufacturer Code:NBP273452AF594
Catalog # NP27342A94
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI5E1 |
Immunogen | Full length recombinant protein of human PKMYT1 (NP_004194) produced in HEK293T cells. |
Conjugate | Alexa Fluor 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: DKFZp547K1610 EC 2.7.11.1 FLJ20093 Myt1 kinase MYT1membrane-associated tyrosine- and threonine-specific cdc2-inhibitory kinase protein kinase membrane associated tyrosine/threonine 1; Membrane-associated tyrosine- and threonine-specific cdc2-inhibitory kinase; Myt1 kinase; protein phosphatase 1, regulatory subunit 126
Gene Aliases: MYT1; PKMYT1; PPP1R126
UniProt ID: (Human) Q99640
Entrez Gene ID: (Human) 9088
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.