Proteintech
Manufacturer Code:256591AP150UL
Catalog # 50-173-5010
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PKM Fusion Protein Ag20270 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CTHBP; CTHBP OIP 3 OIP3 Opa interacting protein 3 p58 PK2 PK3 PKM PKM1 PKM2 PKM2 pyruvate kinase muscle Pyruvate kinase 2/3 Pyruvate kinase isozymes M1/M2 Pyruvate kinase muscle isozyme TCB THBP1 Tumor M2 PK; Cytosolic thyroid hormone-binding protein; epididymis secretory protein Li 30; OIP-3; OPA-interacting protein 3; p58; PK, muscle type; Pyruvate kinase 2/3; pyruvate kinase isozymes M1/M2; Pyruvate kinase muscle isozyme; Pyruvate kinase PKM; THBP1; Thyroid hormone-binding protein 1; thyroid hormone-binding protein, cytosolic; Tumor M2-PK
Gene Aliases: CTHBP; HEL-S-30; OIP3; PK2; PK3; PKM; PKM2; TCB; THBP1
UniProt ID: (Human) B4DUU6
Entrez Gene ID: (Human) 5315
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.