Proteintech
Manufacturer Code:224561AP150UL
Catalog # 50-173-3870
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PKLR Fusion Protein Ag17933 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: L-PK; PK1 PKL PKLR PKR PKRL Pyruvate kinase 1 Pyruvate kinase isozymes R/L pyruvate kinase liver and RBC R type/L type pyruvate kinase Red cell/liver pyruvate kinase RPK; Pyruvate kinase 1; pyruvate kinase isozyme R/L; Pyruvate kinase isozymes L/R; pyruvate kinase isozymes R/L; Pyruvate kinase PKLR; pyruvate kinase type L; pyruvate kinase, liver and blood cell; pyruvate kinase, liver and red blood cell; R-type/L-type pyruvate kinase; Red cell/liver pyruvate kinase
Gene Aliases: Pk-1; PK1; PKL; Pklg; PKLR; PKR; PKRL; R-PK; RPK
UniProt ID: (Human) P30613, (Rat) P12928
Entrez Gene ID: (Human) 5313, (Mouse) 18770, (Rat) 24651
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.