Abnova
Manufacturer Code:PAB1005
Catalog # 89-114-586
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to human PKD2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of PKD2.
For Research Use Only
Protein Aliases: APKD2 MGC138466 MGC138468 PC2 PKD4; Autosomal dominant polycystic kidney disease type II protein; PC2; polycystic kidney disease 2 (autosomal dominant); Polycystic kidney disease 2 protein; Polycystin-2; Polycystwin; R48321; Transient receptor potential cation channel subfamily P member 2; transient receptor potential cation channel, subfamily P, member 2
Gene Aliases: APKD2; Pc-2; PC2; PKD2; PKD4; TRPP2
UniProt ID: (Human) Q13563
Entrez Gene ID: (Human) 5311
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.