Abnova
Manufacturer Code:PAB21493
Catalog # 89-122-986
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human PKD1L1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant PKD1L1.
For Research Use Only
Protein Aliases: PC1-like 1 protein; polycystic kidney disease 1 like 1; Polycystic kidney disease protein 1-like 1; Polycystin-1L1; PRO19563
Gene Aliases: PKD1L1; PRO19563; UNQ5785/PRO19563
UniProt ID: (Human) Q8TDX9
Entrez Gene ID: (Human) 168507
Molecular Function: G-protein modulator enzyme modulator ion channel membrane-bound signaling molecule signaling molecule transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.