Novus Biologicals
Manufacturer Code:NBP267516
Catalog # NBP267516
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ST48-06 |
Immunogen | Recombinant Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
Protein Aliases: EC 2.7.11 EC 2.7.11.13 MGC41878 PKC-B PKC-beta PKCBPRKCB2 PRKCB1protein kinase C beta 1 protein kinase C beta type protein kinase C beta protein kinase C beta 1 protein kinase C beta 1 polypeptide; PKC-B; Protein kinase C beta type; protein kinase C, beta 1 polypeptide
Gene Aliases: PKC-beta; PKCB; PRKCB; PRKCB1; PRKCB2
UniProt ID: (Human) P05771
Entrez Gene ID: (Human) 5579
Molecular Function: annexin calcium-binding protein calmodulin intracellular calcium-sensing protein kinase non-receptor serine/threonine protein kinase protein kinase transfer/carrier protein transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.