R&D Systems
Manufacturer Code:IC4368N100UG
Catalog # IC4368N100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 453416 |
Immunogen | <EM>E. coli</EM>-derived recombinant human PKC theta His580-Ser706 Accession # Q04759 |
Conjugate | Alexa Fluor 700 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Mouse MonoclonalAntibody
For Research Use Only
Protein Aliases: EC 2.7.11 EC 2.7.11.13 MGC126514 MGC141919 nPKC-theta PRKCQ PRKCT protein kinase C theta type protein kinase C theta; nPKC-theta; Protein kinase C theta type; protein kinase C, theta
Gene Aliases: nPKC-theta; PRKCQ; PRKCT
UniProt ID: (Human) Q04759
Entrez Gene ID: (Human) 5588
Molecular Function: annexin calcium-binding protein calmodulin intracellular calcium-sensing protein kinase non-receptor serine/threonine protein kinase protein kinase transfer/carrier protein transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.