Invitrogen
Manufacturer Code:PA570232
Catalog # PIPA570232
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the C-terminal region of mouse PKC theta |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: nPKC-theta; PKC theta; Protein kinase C theta type; Protein kinase C theta type nPKC-theta PRKCQ PRKCT; protein kinase C, theta
Gene Aliases: A130035A12Rik; AW494342; nPKC-theta; PKC-0; PKC-theta; Pkcq; PKCtheta; PRKCQ; PRKCT; si:ch211-239o15.1
UniProt ID: (Human) Q04759, (Mouse) Q02111
Entrez Gene ID: (Dog) 478013, (Bovine) 505901, (Horse) 100070207, (Rabbit) 103350889, (Human) 5588, (Guinea pig) 100726308, (Mouse) 18761, (Rat) 85420, (Zebrafish) 555521
Molecular Function: annexin calcium-binding protein calmodulin intracellular calcium-sensing protein kinase non-receptor serine/threonine protein kinase protein kinase transfer/carrier protein transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.