Invitrogen
Manufacturer Code:MA534757
Catalog # PIMA534757
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | JG69-32 |
Immunogen | Synthetic peptide within Human PKC gamma aa 350-450. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles store in dark |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: DXS1179E MGC57564 nPKC-delta nPKC-epsilon nPKC-eta nPKC-theta nPKC-zeta PKC PKC2 PKCA PKC-A PKC-alpha PKCB PKC-B PKC-beta Pkcc PKCE Pkcg PKCgamma PKC-gamma PKCI PKCL PKC-L PRKACA Prkc PRKC Pan PRKCA PRKCB1 Prkcc PRKCD PRKCE Prkcg PRKCH PRKCI PRKCL PRKCQ PRKCT PRKCZ Protein kinase C gamma type RATPKCI SCA14 SDK-1; PKC-gamma; Protein kinase C gamma type; protein kinase C, gamma
Gene Aliases: PKC-gamma; PKCC; PKCG; PRKCG; SCA14
UniProt ID: (Human) P05129
Entrez Gene ID: (Human) 5582
Molecular Function:
annexin
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
kinase
non-receptor serine/threonine protein kinase
protein kinase
transfer/carrier protein
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.