Proteintech
Manufacturer Code:664291IG150UL
Catalog # 50-173-6856
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2F4B9 |
Immunogen | PKC gamma Fusion Protein Ag5910 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: PKC gamma PKCC PKCG PRKCG Protein kinase C gamma type protein kinase C gamma SCA14; PKC-gamma; Protein kinase C gamma type; protein kinase C type I (gamma type); protein kinase C, gamma
Gene Aliases: PKC; PKC-gamma; PKCC; PKCG; PKCgamma; PKCI; Prkc; Prkcc; PRKCG; RATPKCI; SCA14
UniProt ID: (Human) P05129, (Mouse) P63318, (Rat) P63319
Entrez Gene ID: (Human) 5582, (Pig) 100518074, (Mouse) 18752, (Rat) 24681
Molecular Function: annexin calcium-binding protein calmodulin intracellular calcium-sensing protein kinase non-receptor serine/threonine protein kinase protein kinase transfer/carrier protein transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.