Invitrogen
Manufacturer Code:PA586906
Catalog # PIPA586906
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 543-587 of Human PRKCG |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: PKC-gamma; Protein kinase C gamma type; Protein kinase C gamma type PKC-gamma; protein kinase C type I (gamma type); protein kinase C, gamma
Gene Aliases: PKC; PKC-gamma; PKCC; PKCG; PKCgamma; PKCI; Prkc; Prkcc; PRKCG; RATPKCI; SCA14
UniProt ID: (Human) P05129, (Mouse) P63318, (Rat) P63319
Entrez Gene ID: (Human) 5582, (Mouse) 18752, (Rat) 24681
Molecular Function:
annexin
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
kinase
non-receptor serine/threonine protein kinase
protein kinase
transfer/carrier protein
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.