Invitrogen
Manufacturer Code:PA513747
Catalog # PIPA513747
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 305-336 amino acids from the C-terminal region of human PKC gamma |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: PKC-gamma; Protein kinase C gamma type; Protein kinase C gamma type PKC-gamma; protein kinase C, gamma
Gene Aliases: PKC-gamma; PKCC; PKCG; PKCgamma; Prkcc; PRKCG; SCA14
UniProt ID: (Human) P05129, (Mouse) P63318
Entrez Gene ID: (Human) 5582, (Mouse) 18752
Molecular Function: annexin calcium-binding protein calmodulin intracellular calcium-sensing protein kinase non-receptor serine/threonine protein kinase protein kinase transfer/carrier protein transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.