Invitrogen
Manufacturer Code:LFPA0174
Catalog # PILFPA0174
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MGC57564 PKC-gamma PKCC PKCG SCA14; PKC-gamma; Protein kinase C gamma type; protein kinase C type I (gamma type)
Gene Aliases: PKC; Pkcc; Pkcg; PKCgamma; PKCI; Prkc; Prkcc; Prkcg; RATPKCI
UniProt ID: (Mouse) P63318, (Rat) P63319
Entrez Gene ID: (Mouse) 18752, (Rat) 24681
Molecular Function:
annexin
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
kinase
non-receptor serine/threonine protein kinase
protein kinase
transfer/carrier protein
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.