R&D Systems
Manufacturer Code:MAB51341100
Catalog # MAB51341100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 666843 |
Immunogen | <em>E. coli</em>-derived recombinant human PKC epsilon Gln580-Pro737 Accession # Q02156 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: EC 2.7.11 EC 2.7.11.13 MGC125656 nPKC-epsilon PKCE PKCEMGC125657 PRKCE protein kinase C epsilon type protein kinase C epsilon; nPKC-epsilon; Protein kinase C epsilon type; protein kinase C, epsilon
Gene Aliases: nPKC-epsilon; PKCE; PRKCE
UniProt ID: (Human) Q02156
Entrez Gene ID: (Human) 5581
Molecular Function: annexin calcium-binding protein calmodulin intracellular calcium-sensing protein kinase non-receptor serine/threonine protein kinase protein kinase transfer/carrier protein transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.