Novus Biologicals
Manufacturer Code:NBP190351
Catalog # NBP190351
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:ANLCGINQKLLAEALNQVTQRASRRSDSASSEPVGIYQGFEKKTGVAGEDMQDNSGTYGKIWEGSSKCNINNFIFHKVLGKGSFGKVLLGELKGRGEYFAIKALKKDVVLIDDDVECTMVEKRVLTLAAENPFLTHLICTFQTKDH |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.7.11 EC 2.7.11.13 MAY1 MGC49908 nPKC-delta PKCD PKCy protein kinase C delta type protein kinase C delta VIII protein kinase C delta; nPKC-delta; Protein kinase C delta type; Protein kinase C delta type catalytic subunit; Protein kinase C delta type regulatory subunit; protein kinase C delta VIII; protein kinase C, delta; SDK1; Sphingosine-dependent protein kinase-1; Tyrosine-protein kinase PRKCD
Gene Aliases: ALPS3; CVID9; MAY1; nPKC-delta; PKCD; PRKCD
UniProt ID: (Human) Q05655
Entrez Gene ID: (Human) 5580
Molecular Function: annexin calcium-binding protein calmodulin intracellular calcium-sensing protein kinase non-receptor serine/threonine protein kinase protein kinase transfer/carrier protein transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.