Novus Biologicals
Manufacturer Code:NBP266799
Catalog # NBP266799
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | JF0964 |
Immunogen | Synthetic phospho-peptide corresponding to residues surrounding Thr638 of human PKC alpha |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
Protein Aliases: AAG6 aging-associated gene 6 EC 2.7.11 EC 2.7.11.13 MGC129901 PKC-A PKC-alpha PKCAMGC129900 PRKACA protein kinase C alpha type protein kinase C alpha; aging-associated gene 6; PKC-A; Protein kinase C alpha type; protein kinase C, alpha
Gene Aliases: AAG6; PKC-alpha; PKCA; PRKACA; PRKCA
UniProt ID: (Human) P17252
Entrez Gene ID: (Human) 5578
Molecular Function:
annexin
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
kinase
non-receptor serine/threonine protein kinase
protein kinase
transfer/carrier protein
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.