Invitrogen
Manufacturer Code:PA585652
Catalog # PIPA585652
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human PKC alpha. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: aging-associated gene 6; PKC-A; PKC-alpha; protein kinase C alpha Antibody PRKCA Antibody AAG6 Antibody PKC-alpha Antibody PKCA Antibody PRKACA Antibody; Protein kinase C alpha type; protein kinase C, alpha; protein kinase c-alpha
Gene Aliases: AAG6; AI875142; PKC-alpha; PKCA; PRKACA; PRKCA
UniProt ID: (Human) P17252, (Mouse) P20444
Entrez Gene ID: (Human) 5578, (Mouse) 18750, (Rat) 24680
Molecular Function:
annexin
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
kinase
non-receptor serine/threonine protein kinase
protein kinase
transfer/carrier protein
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.