Proteintech
Manufacturer Code:156591AP150UL
Catalog # 50-173-0382
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PIWIL1 Fusion Protein Ag8056 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HIWI MIWI PIWI piwi like 1 (Drosophila) Piwi like protein 1 PIWIL1; piwi homolog; piwi like homolog 1; piwi-like homolog 1; Piwi-like protein 1; piwi-like RNA-mediated gene silencing 1
Gene Aliases: CT80.1; HIWI; MIWI; PIWI; PIWIL1
UniProt ID: (Human) Q96J94, (Mouse) Q9JMB7
Entrez Gene ID: (Human) 9271, (Mouse) 57749
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.