Invitrogen
Manufacturer Code:PA595968
Catalog # PIPA595968
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence within amino acids 1-100 of human PITX2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: all1-responsive gene 1; ALL1-responsive protein ARP1; BRX1 homeoprotein; Homeobox protein PITX2; homeodomain transcription factor 2; Orthodenticle-like homeobox 2; paired-like homeodomain 2; Paired-like homeodomain transcription factor 2; Paired-like homeodomain transcription factor Munc 30; Pituitary homeobox 2; RIEG bicoid-related homeobox transcription factor; rieg bicoid-related homeobox transcription factor 1; rPtx2; Solurshin
Gene Aliases: 9430085M16Rik; ARP1; Brx1; Brx1b; IDG2; IGDS; IGDS2; IHG2; IRID2; Munc30; Otlx2; Pitx-2; PITX2; Pitx2c; PTX2; RGS; RIEG; RIEG1; RS
UniProt ID: (Human) Q99697, (Rat) Q9R0W1, (Mouse) P97474
Entrez Gene ID: (Human) 5308, (Rat) 54284, (Mouse) 18741
Molecular Function: DNA binding protein helix-turn-helix transcription factor homeobox transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.