Invitrogen
Manufacturer Code:PA585500
Catalog # PIPA585500
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the center region of human PITX2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: all1-responsive gene 1; ALL1-responsive protein ARP1; ARP1 antibody Brx1 antibody IDG2 antibody IGDS antibody IGDS2 antibody IHG2 antibody IRID2 antibody MGC111022 antibody MGC20144 antibody Otlx2 antibody PTX2 antibody RGS antibody RIEG antibody RIEG1 antibody RS antibody PITX2 antibody solurshin antibody homeobox protein PITX2 antibody paired-like homeodomain transcription factor 2 antibody ALL1-responsive protein ARP1 antibody pituitary homeo box 2 antibody rieg bicoid-related homeobox transcription factor 1 antibody pituitary homeobox 2 antibody all1-responsive gene 1 antibody paired-like homeodomain 2 antibody; Homeobox protein PITX2; paired-like homeodomain 2; Paired-like homeodomain transcription factor 2; Pituitary homeobox 2; RIEG bicoid-related homeobox transcription factor; rieg bicoid-related homeobox transcription factor 1; Solurshin
Gene Aliases: ARP1; Brx1; IDG2; IGDS; IGDS2; IHG2; IRID2; Otlx2; PITX2; PTX2; RGS; RIEG; RIEG1; RS
UniProt ID: (Human) Q99697
Entrez Gene ID: (Human) 5308
Molecular Function:
DNA binding protein
helix-turn-helix transcription factor
homeobox transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.