Invitrogen
Manufacturer Code:PA565596
Catalog # PIPA565596
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human PITX2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: all1-responsive gene 1; ALL1-responsive protein ARP1; Homeobox protein PITX2; paired-like homeodomain 2; Paired-like homeodomain transcription factor 2; Pituitary homeobox 2; Pituitary homeobox 2 RIEG bicoid-related homeobox transcription factor Solurshin ALL1-responsive protein ARP1 PITX2 ARP1 RGS RIEG RIEG1; RIEG bicoid-related homeobox transcription factor; rieg bicoid-related homeobox transcription factor 1; Solurshin
Gene Aliases: ARP1; Brx1; IDG2; IGDS; IGDS2; IHG2; IRID2; Otlx2; PITX2; PTX2; RGS; RIEG; RIEG1; RS
UniProt ID: (Human) Q99697
Entrez Gene ID: (Human) 5308
Molecular Function: DNA binding protein helix-turn-helix transcription factor homeobox transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.