Abnova
Manufacturer Code:H00005308K
Catalog # 89-200-134
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human PITX2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human PITX2 peptide using ARM Technology.
For Research Use Only
Protein Aliases: all1-responsive gene 1; ALL1-responsive protein ARP1; ARP1 Brx1 IDG2 IGDS IGDS2 IHG2 IRID2 MGC111022 MGC20144 Otlx2 PTX2 RGS RIEG RIEG1 RS; Homeobox protein PITX2; paired-like homeodomain 2; Paired-like homeodomain transcription factor 2; Pituitary homeobox 2; RIEG bicoid-related homeobox transcription factor; rieg bicoid-related homeobox transcription factor 1; Solurshin
Gene Aliases: ARP1; Brx1; IDG2; IGDS; IGDS2; IHG2; IRID2; Otlx2; PITX2; PTX2; RGS; RIEG; RIEG1; RS
UniProt ID: (Human) Q99697
Entrez Gene ID: (Human) 5308
Molecular Function: DNA binding protein helix-turn-helix transcription factor homeobox transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.