Invitrogen
Manufacturer Code:PA541438
Catalog # PIPA541438
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human PITX1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hindlimb expressed homeobox protein backfoot; Hindlimb-expressed homeobox protein backfoot; Homeobox protein P-OTX; Homeobox protein PITX1; paired-like homeodomain 1; Paired-like homeodomain 1 BFT CCF POTX PTX1; Paired-like homeodomain transcription factor 1; pituitary homeo box 1; Pituitary homeobox 1; Pituitary OTX-related factor
Gene Aliases: BFT; CCF; LBNBG; P-OTX; PITX1; POTX; PTX1; zgc:136976
UniProt ID: (Human) P78337, (Mouse) P70314, (Rat) Q99NA7
Entrez Gene ID: (Horse) 100072706, (Human) 5307, (Dog) 481513, (Bovine) 508754, (Mouse) 18740, (Guinea pig) 100715612, (Rat) 113983, (Zebrafish) 678598
Molecular Function:
DNA binding protein
helix-turn-helix transcription factor
homeobox transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.