Invitrogen
Manufacturer Code:PA5101136
Catalog # PIPA5101136
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human PITX1 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BFT CCF hindlimb expressed homeobox protein backfoot hindlimb-expressed homeobox protein backfoot homeobox protein PITX1 Homeobox protein P-OTX LBNBG OTX paired like homeodomain 1 paired-like homeodomain 1 paired-like homeodomain transcription factor 1 pituitary homeo box 1 pituitary homeobox 1 Pituitary OTX-related factor PITX1 Potx P-OTX PTX1; hindlimb expressed homeobox protein backfoot; Hindlimb-expressed homeobox protein backfoot; Homeobox protein P-OTX; Homeobox protein PITX1; paired-like homeodomain 1; Paired-like homeodomain transcription factor 1; pituitary homeo box 1; Pituitary homeobox 1; Pituitary OTX-related factor
Gene Aliases: BFT; CCF; LBNBG; P-OTX; PITX1; POTX; PTX1
UniProt ID: (Human) P78337, (Mouse) P70314, (Rat) Q99NA7
Entrez Gene ID: (Human) 5307, (Mouse) 18740, (Rat) 113983
Molecular Function:
DNA binding protein
helix-turn-helix transcription factor
homeobox transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.