Invitrogen
Manufacturer Code:PA521903
Catalog # PIPA521903
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to a region within amino acids 912 and 974 of NIR1 (Uniprot IDNo.Q9BZ71) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: atypical chemokine receptor 6; cone rod dystrophy 5; CORD5 NIR1 RDGBA3 NIR-1 PITPnm 3 PYK2 N-terminal domain-interacting receptor 1 membrane-associated phosphatidylinositol transfer protein 3 phosphatidylinositol transfer protein membrane-associated 3 retinal degeneration B alpha 3; Membrane-associated phosphatidylinositol transfer protein 3; NIR-1; Phosphatidylinositol transfer protein, membrane-associated 3; PITPnm 3; PYK2 N-terminal domain-interacting receptor 1; retinal degeneration B alpha 3
Gene Aliases: ACKR6; CORD5; NIR1; PITPNM3; RDGBA3
UniProt ID: (Human) Q9BZ71
Entrez Gene ID: (Human) 83394
Molecular Function:
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.