Novus Biologicals
Manufacturer Code:NBP131070
Catalog # NBP131070
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human PITPNM3. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: atypical chemokine receptor 6; cone rod dystrophy 5; Membrane-associated phosphatidylinositol transfer protein 3; membrane-associated phosphatidylinositol transfer protein 3 MGC157740 MGC157741 NIR-1 NIR1CORD5 Phosphatidylinositol transfer protein membrane-associated 3 PITPnm 3 PITPNM family member 3 Pyk2 N-terminal domain-interacting receptor 1 RDGBA3 retinal degeneration B alpha 3; NIR-1; Phosphatidylinositol transfer protein, membrane-associated 3; PITPnm 3; PYK2 N-terminal domain-interacting receptor 1; retinal degeneration B alpha 3
Gene Aliases: ACKR6; CORD5; NIR1; PITPNM3; RDGBA3
UniProt ID: (Human) Q9BZ71
Entrez Gene ID: (Human) 83394
Molecular Function: transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.