Novus Biologicals
Manufacturer Code:NB300814
Catalog # NB300814
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-SWARGPPKFESVP corresponding to C-Terminus according to NP_112497.2 NP_001159438.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: atypical chemokine receptor 6; cone rod dystrophy 5; Membrane-associated phosphatidylinositol transfer protein 3; membrane-associated phosphatidylinositol transfer protein 3 MGC157740 MGC157741 NIR-1 NIR1CORD5 Phosphatidylinositol transfer protein membrane-associated 3 PITPnm 3 PITPNM family member 3 Pyk2 N-terminal domain-interacting receptor 1 RDGBA3 retinal degeneration B alpha 3; NIR-1; Phosphatidylinositol transfer protein, membrane-associated 3; PITPnm 3; PYK2 N-terminal domain-interacting receptor 1; retinal degeneration B alpha 3
Gene Aliases: ACKR6; CORD5; NIR1; PITPNM3; RDGBA3
UniProt ID: (Human) Q9BZ71
Entrez Gene ID: (Human) 83394
Molecular Function: transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.