Invitrogen
Manufacturer Code:PA563823
Catalog # PIPA563823
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human PITPNM1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Drosophila retinal degeneration B homolog; Membrane-associated phosphatidylinositol transfer protein 1; NIR-2; phosphatidylinositol transfer protein, membrane associated 1; Phosphatidylinositol transfer protein, membrane-associated 1; PITPnm 1; PITPNM1 PITPNM NIR2 DRES9 phosphatidylinositol transfer protein membrane-associated Drosophila retinal degeneration B PYK2 N-terminal domain-interacting receptor 2 phosphatidylinositol transfer protein membrane-associated 1 FLJ44997 RDGB RDGB1 RDGBA RDGBA1 Rd9 phosphatidylinositol transfer protein membrane-associated retinal degeneration B alpha 1; PYK2 N-terminal domain-interacting receptor 2; retinal degeneration B alpha 1
Gene Aliases: DRES9; NIR2; PITPNM; PITPNM1; Rd9; RDGB; RDGB1; RDGBA; RDGBA1
UniProt ID: (Human) Q9BZ73
Entrez Gene ID: (Human) 9600
Molecular Function:
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.