Invitrogen
Manufacturer Code:PA530508
Catalog # PIPA530508
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 219 of Human PITPNC1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cytoplasmic phosphatidylinositol transfer protein 1; M-rdgB beta; Mammalian rdgB homolog beta; RDGB-BETA RDGBB RDGBB1 M-rdgB beta cytoplasmic phosphatidylinositol transfer protein 1 mammalian rdgB homolog beta mrdgBbeta rdgBbeta retinal degeneration B beta 1 retinal degeneration B homolog beta; RdgBbeta; retinal degeneration B beta 1; Retinal degeneration B homolog beta
Gene Aliases: M-RDGB-beta; MRDGBbeta; PITPNC1; RDGB-BETA; RDGBB; RDGBB1
UniProt ID: (Human) Q9UKF7
Entrez Gene ID: (Human) 26207
Molecular Function:
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.