Proteintech
Manufacturer Code:131101AP150UL
Catalog # 50-172-8495
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PITPNB Fusion Protein Ag3774 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Phosphatidylinositol transfer protein beta isoform; phosphatidylinositol transfer protein, beta; phosphotidylinositol transfer protein, beta; PI TP beta PITPNB PtdIns transfer protein beta PtdInsTP PtdInsTP beta VIB1B; PI-TP-beta; Pitp beta; PtdIns transfer protein beta; ptdInsTP beta
Gene Aliases: PI-TP-beta; PITPNB; PtdInsTP; VIB1B
UniProt ID: (Human) P48739, (Rat) P53812, (Mouse) P53811
Entrez Gene ID: (Human) 23760, (Rat) 114561, (Mouse) 56305
Molecular Function:
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.