Invitrogen
Manufacturer Code:PA19083
Catalog # 01-674-667
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to the C terminus amino acids RQKDPVKGMTADD. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: epididymis secretory protein Li 36; Phosphatidylinositol transfer protein alpha isoform; phosphatidylinositol transfer protein, alpha; PI-TP-alpha; PITPN PITPNB phosphotidylinositol transfer protein phosphatidylinositol transfer protein PITP alpha; ptdIns transfer protein alpha; ptdInsTP alpha
Gene Aliases: HEL-S-36; PI-TPalpha; PITPN; PITPNA; VIB1A
UniProt ID: (Human) Q00169
Entrez Gene ID: (Human) 5306
Molecular Function: transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.