Invitrogen
Manufacturer Code:PA124436
Catalog # PIPA124436
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide region of human POU1F1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GHF-1; Growth hormone factor 1; homeodomain protein for Growth Hormone (GH); PIT-1; Pituary specific transcription factor 1 (growth hormone factor 1); pituitary specific transcription factor 1; pituitary specific transcription factor 1, related sequence 1; Pituitary-specific positive transcription factor 1; pituitary-specific positive transcription factor 1 variant w; POU domain, class 1, transcription factor 1; POU domain, class 1, transcription factor 1 (Pit1, growth hormone factor 1); POU1F1
Gene Aliases: CPHD1; dw; dwarf; GHF-1; GHF1; GHF1A; Hmp1; PIT-1; PIT1; Pit1-rs1; PIT1Z; POU1F1; POU1F1a
UniProt ID: (Dog) Q9TTI7, (Human) P28069, (Mouse) Q00286, (Rat) P10037
Entrez Gene ID: (Dog) 403753, (Human) 5449, (Mouse) 18736, (Rat) 25517
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.