Novus Biologicals
Manufacturer Code:NBP275618
Catalog # NBP275618
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | JG79-35 |
Immunogen | Recombinant protein within human PIST aa 1-100. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
Protein Aliases: CALPDZ protein interacting specifically with TC10 CFTR-associated ligand dJ94G16.2 FIGdJ94G16.2 PIST Fused in glioblastoma golgi-associated PDZ and coiled-coil motif containing Golgi-associated PDZ and coiled-coil motif-containing protein GOPC1 PDZ/coiled-coil domain binding partner for the rho-family GTPase TC10 PISTGolgi associated PDZ and coiled-coil motif containing protein; CFTR-associated ligand; dJ94G16.2 PIST; Fused in glioblastoma; Golgi associated PDZ and coiled-coil motif containing protein; golgi-associated PDZ and coiled-coil motif containing protein; Golgi-associated PDZ and coiled-coil motif-containing protein; PDZ protein interacting specifically with TC10; PDZ/coiled-coil domain binding partner for the rho-family GTPase TC10; PIST
Gene Aliases: CAL; dJ94G16.2; FIG; GOPC; GOPC1; PIST
UniProt ID: (Human) Q9HD26
Entrez Gene ID: (Human) 57120
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.