Proteintech
Manufacturer Code:209901AP150UL
Catalog # 50-173-3194
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hCG&lowbar1776018 PIRT TRPV1 complex; Phosphoinositide-interacting protein; phosphoinositide-interacting regulator of transient receptor potential channels; phosphoinositide-interacting regulator of TRPV1
Gene Aliases: PIRT
UniProt ID: (Human) P0C851
Entrez Gene ID: (Human) 644139
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.