Novus Biologicals
Manufacturer Code:NBP273417R
Catalog # NBP273417R
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3D3 |
Immunogen | Full length recombinant protein of human PIP5K2 alpha (NP_005019) produced in HEK293T cells. |
Conjugate | DyLight 550 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: 1-phosphatidylinositol 5-phosphate 4-kinase 2-alpha; 1-phosphatidylinositol-4-phosphate kinase; 1-phosphatidylinositol-4-phosphate kinase 1-phosphatidylinositol-4-phosphate-5-kinase 1-phosphatidylinositol-5-phosphate 4-kinase 2-alpha Diphosphoinositide kinase 2-alpha EC 2.7.1 EC 2.7.1.149 FLJ13267 phosphatidylinositol-4-phosphate 5-kinase type II alpha Phosphatidylinositol-5-phosphate 4-kinase type II alpha phosphatidylinositol-5-phosphate 4-kinase type-2 alpha phosphatidylinositol-5-phosphate 4-kinase type II alpha PI(5)P 4-kinase type II alpha PIP4KII-alpha PIP5K2 PIP5K2API5P4KA PIP5KIIA PIP5KIIalpha PIP5KII-alpha PIP5KIII PIPK PtdIns(4)P-5-kinase B isoform PtdIns(4)P-5-kinase C isoform PtdIns(5)P-4-kinase isoform 2-alpha type II phosphatidylinositol-4-phosphate 5-kinase 53 K isoform; 1-phosphatidylinositol-4-phosphate-5-kinase; 1-phosphatidylinositol-5-phosphate 4-kinase 2-alpha; Diphosphoinositide kinase 2-alpha; Phosphatidylinositol 5-Phosphate 4-Kinase; Phosphatidylinositol 5-phosphate 4-kinase type II alpha; Phosphatidylinositol 5-phosphate 4-kinase type-2 alpha; phosphatidylinositol-4-phosphate 5-kinase, type II, alpha; phosphatidylinositol-5-phosphate 4-kinase type-2 alpha; phosphatidylinositol-5-phosphate 4-kinase, type II, alpha; PI(5)P 4-kinase type II alpha; PI5P4Kalpha; PIP4KII-alpha; PIP5KIIalpha; PIP5KIII; PtdIns(4)P-5-kinase B isoform; PtdIns(4)P-5-kinase C isoform; PtdIns(5)P-4-kinase isoform 2-alpha; type II phosphatidylinositol-4-phosphate 5-kinase 53 K isoform
Gene Aliases: PI5P4KA; PIP4K2A; PIP5K2; PIP5K2A; PIP5KII-alpha; PIP5KIIA; PIPK
UniProt ID: (Human) P48426
Entrez Gene ID: (Human) 5305
Molecular Function:
kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.