Invitrogen
Manufacturer Code:PA528274
Catalog # PIPA528274
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 152 and 375 of Human PIP5K1C |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: diphosphoinositide kinase; LCCS3 PIP5K-GAMMA PIP5Kgamma PIPKIg&lowbarv4 Human homolog of mouse phosphatidylinositol-4-phosphate 5-kinase I-gamma PIP5K1-gamma diphosphoinositide kinase phosphatidylinositol-4-phosphate 5-kinase type-1 gamma ptdIns(4)P-5-kinase 1 gamma type I PIP kinase; Phosphatidylinositol 4-phosphate 5-kinase type I gamma; Phosphatidylinositol 4-phosphate 5-kinase type-1 gamma; phosphatidylinositol-4-phosphate 5-kinase type I gamma; phosphatidylinositol-4-phosphate 5-kinase type-1 gamma; phosphatidylinositol-4-phosphate 5-kinase, type I, gamma; PIP5K1-gamma; PIP5K1gamma; PIP5KIgamma; ptdIns(4)P-5-kinase 1 gamma; Type I phosphatidylinositol 4-phosphate 5-kinase gamma; type I PIP kinase
Gene Aliases: AI115456; AI835305; KIAA0589; LCCS3; PIP5K-GAMMA; PIP5K1-gamma; PIP5K1C; PIP5Kgamma; Pip5kIgamma
UniProt ID: (Human) O60331, (Mouse) O70161
Entrez Gene ID: (Human) 23396, (Mouse) 18717
Molecular Function: kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.