Invitrogen
Manufacturer Code:PA5115760
Catalog # PIPA5115760
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human PIP5K1C corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AI115456 AI835305 diphosphoinositide kinase KIAA0589 LCCS3 Phosphatidylinositol 4-phosphate 5-kinase type I gamma phosphatidylinositol 4-phosphate 5-kinase type-1 gamma phosphatidylinositol-4-phosphate 5-kinase phosphatidylinositol-4-phosphate 5-kinase type 1 gamma phosphatidylinositol-4-phosphate 5-kinase type I gamma phosphatidylinositol-4-phosphate 5-kinase type-1 gamma phosphatidylinositol-4-phosphate 5-kinase type 1 gamma phosphatidylinositol-4-phosphate 5-kinase type I gamma PIP5K1C PIP5K1-gamma PIP5Kgamma PIP5K-GAMMA PIP5KI gamma Pip5kIgamma PIP5KI-gamma PIPKIg_v4 ptdIns(4)P-5-kinase 1 gamma ptdIns(4)P-5-kinase gamma ptdInsPKIgamma type I PIP kinase; diphosphoinositide kinase; Phosphatidylinositol 4-phosphate 5-kinase type-1 gamma; phosphatidylinositol-4-phosphate 5-kinase, type I, gamma; PIP5K1gamma; ptdIns(4)P-5-kinase 1 gamma; Type I phosphatidylinositol 4-phosphate 5-kinase gamma; type I PIP kinase
Gene Aliases: KIAA0589; LCCS3; PIP5K-GAMMA; PIP5K1-gamma; PIP5K1C; PIP5Kgamma
UniProt ID: (Human) O60331
Entrez Gene ID: (Human) 23396
Molecular Function: kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.