Novus Biologicals
Manufacturer Code:NBP182986
Catalog # NBP182986
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:RPSFYAERFFKFMSNTVFRKNSSLKSSPSKKGRGGALLAVKPLGPTAAFSASQIPSEREEAQYDLRGARSYPTLEDEGRPDLLPCTPPSFEEATTASIATTLSSTSLSIPERS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: diphosphoinositide kinase; EC 2.7.1 EC 2.7.1.68 KIAA0589diphosphoinositide kinase LCCS3Human homolog of mouse phosphatidylinositol-4-phosphate 5-kinase I-gamma Phosphatidylinositol-4-phosphate 5-kinase type I gamma phosphatidylinositol-4-phosphate 5-kinase type-1 gamma phosphatidylinositol-4-phosphate 5-kinase type I gamma PIP5K1-gamma PIP5Kgamma PIP5K-GAMMA PIP5KIgamma PIPKIg_v4 PtdIns(4)P-5-kinase 1 gamma type I PIP kinase; Phosphatidylinositol 4-phosphate 5-kinase type-1 gamma; phosphatidylinositol-4-phosphate 5-kinase, type I, gamma; PIP5K1gamma; ptdIns(4)P-5-kinase 1 gamma; Type I phosphatidylinositol 4-phosphate 5-kinase gamma; type I PIP kinase
Gene Aliases: KIAA0589; LCCS3; PIP5K-GAMMA; PIP5K1-gamma; PIP5K1C; PIP5Kgamma
UniProt ID: (Human) O60331
Entrez Gene ID: (Human) 23396
Molecular Function:
kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.