Invitrogen
Manufacturer Code:PA5115759
Catalog # PIPA5115759
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human PIP5K1B corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 68 kDa type I phosphatidylinositol 4-phosphate 5-kinase beta; MSS4 Phosphatidylinositol 4-phosphate 5-kinase type I alpha Phosphatidylinositol 4-phosphate 5-kinase type I beta phosphatidylinositol 4-phosphate 5-kinase type-1 beta phosphatidylinositol-4-phosphate 5-kinase phosphatidylinositol-4-phosphate 5-kinase type 1 beta phosphatidylinositol-4-phosphate 5-kinase type I alpha phosphatidylinositol-4-phosphate 5-kinase type I beta phosphatidylinositol-4-phosphate 5-kinase type-1 beta phosphatidylinositol-4-phosphate 5-kinase type 1 alpha phosphatidylinositol-4-phosphate 5-kinase type 1 beta phosphatidylinositol-4-phosphate 5-kinase type 1 beta phosphatidylinositol-4-phosphate 5-kinase type I beta PI4P5K-I[b] Pip5k1a Pip5k1b PIP5K1-beta PIP5KIalpha PIP5KIB2 protein PIP5KIbeta Pipk5b Protein STM-7 ptdIns(4)P-5-kinase 1 beta ptdIns(4)P-5-kinase beta STM7 STM-7 protein Type I phosphatidylinositol 4-phosphate 5-kinase beta type I phosphatidylinositol-4-phosphate 5-kinase beta; Phosphatidylinositol 4-phosphate 5-kinase type I alpha; Phosphatidylinositol 4-phosphate 5-kinase type I beta; Phosphatidylinositol 4-phosphate 5-kinase type-1 beta; phosphatidylinositol-4-phosphate 5-kinase type I alpha; phosphatidylinositol-4-phosphate 5-kinase type I beta; phosphatidylinositol-4-phosphate 5-kinase type-1 beta; phosphatidylinositol-4-phosphate 5-kinase, type 1 alpha; phosphatidylinositol-4-phosphate 5-kinase, type 1, beta; phosphatidylinositol-4-phosphate 5-kinase, type I, beta; PIP5K1-beta; PIP5KI-beta; PIP5KIalpha; PIP5KIbeta; Protein STM-7; ptdIns(4)P-5-kinase 1 beta; ptdIns(4)P-5-kinase beta; Type I phosphatidylinositol 4-phosphate 5-kinase beta; type I phosphatidylinositol-4-phosphate 5-kinase beta
Gene Aliases: MSS4; PI4P5K-I[b]; PIP5K1-beta; Pip5k1a; PIP5K1B; PIP5KIbeta; Pipk5b; STM7
UniProt ID: (Human) B4DIG7, (Mouse) P70181, (Rat) Q5CZZ9
Entrez Gene ID: (Human) 8395, (Mouse) 18719, (Rat) 309419
Molecular Function: kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.