Abnova
Manufacturer Code:PAB3237
Catalog # 89-130-908
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human PIP4K2C. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of PIP4K2C.
For Research Use Only
Protein Aliases: FLJ22055; phosphatidylinositol 4-phosphate 5-kinase; Phosphatidylinositol 5-phosphate 4-kinase type II gamma; Phosphatidylinositol 5-phosphate 4-kinase type-2 gamma; phosphatidylinositol-4-phosphate 5-kinase, type II, gamma; phosphatidylinositol-5-phosphate 4-kinase type-2 gamma; phosphatidylinositol-5-phosphate 4-kinase, type II, gamma; PI(5)P 4-kinase type II gamma; PIP4KII-gamma; PIP5K2C
Gene Aliases: PIP4K2C; PIP5K2C
UniProt ID: (Human) Q8TBX8
Entrez Gene ID: (Human) 79837
Molecular Function: kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.