Proteintech
Manufacturer Code:170771AP150UL
Catalog # 50-173-1370
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PIP4K2C Fusion Protein Ag10989 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: phosphatidylinositol 4-phosphate 5-kinase; Phosphatidylinositol 5-phosphate 4-kinase type II gamma; Phosphatidylinositol 5-phosphate 4-kinase type-2 gamma; phosphatidylinositol-4-phosphate 5-kinase, type II, gamma; phosphatidylinositol-5-phosphate 4-kinase type-2 gamma; phosphatidylinositol-5-phosphate 4-kinase, type II, gamma; PI(5)P 4 kinase type II gamma PIP4K2C PIP4KII gamma PIP5K2C; PI(5)P 4-kinase type II gamma; PIP4KII-gamma
Gene Aliases: PIP4K2C; PIP5K2C
UniProt ID: (Human) Q8TBX8, (Mouse) Q91XU3, (Rat) O88370
Entrez Gene ID: (Human) 79837, (Mouse) 117150, (Rat) 140607
Molecular Function:
kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.