Invitrogen
Manufacturer Code:PA5110172
Catalog # PIPA5110172
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-270 of human PIP4K2B (NP_003550.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1-phosphatidylinositol 5-phosphate 4-kinase 2-beta; 1-phosphatidylinositol 5-phosphate 4-kinase 2-beta AI848124 BOS_19028 c11 Diphosphoinositide kinase 2-beta Phosphatidylinositol 5-phosphate 4-kinase type II beta Phosphatidylinositol 5-phosphate 4-kinase type-2 beta Phosphatidylinositol-phosphate kinase IIgamma PI(5)P 4-kinase type II beta PI5P4KB PI5P4Kbeta Pip4k2b Pip5k2b PIP5KIIB PIP5KIIbeta PIP5P4KB PIPKIIgamma PtdIns(5)P-4-kinase isoform 2-beta; 1-phosphatidylinositol-4-phosphate kinase; 1-phosphatidylinositol-5-phosphate 4-kinase 2-beta; Diphosphoinositide kinase 2-beta; Phosphatidylinositol 5-phosphate 4-kinase type II beta; Phosphatidylinositol 5-phosphate 4-kinase type-2 beta; phosphatidylinositol-4-phosphate 5-kinase, type II, beta; phosphatidylinositol-5-phosphate 4-kinase type-2 beta; phosphatidylinositol-5-phosphate 4-kinase, type II, beta; PI(5)P 4-kinase type II beta; PIP4KII-beta; PTDINS(4)P-5-kinase; PtdIns(5)P-4-kinase isoform 2-beta
Gene Aliases: AI848124; c11; PI5P4KB; PI5P4Kbeta; PIP4K2B; PIP5K2B; PIP5KIIB; PIP5KIIbeta; PIP5P4KB
UniProt ID: (Human) P78356, (Mouse) Q80XI4
Entrez Gene ID: (Human) 8396, (Mouse) 108083
Molecular Function: kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.