Invitrogen
Manufacturer Code:PA531141
Catalog # PIPA531141
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 3 and 233 of Human PIP5K2 alpha |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1-phosphatidylinositol 5-phosphate 4-kinase 2-alpha; 1-phosphatidylinositol-4-phosphate kinase; 1-phosphatidylinositol-4-phosphate-5-kinase; 1-phosphatidylinositol-5-phosphate 4-kinase 2-alpha; 1-phosphatidylinositol-5-phosphate 4-kinase A; Diphosphoinositide kinase 2-alpha; Phosphatidylinositol 5-Phosphate 4-Kinase; Phosphatidylinositol 5-phosphate 4-kinase type II alpha; Phosphatidylinositol 5-phosphate 4-kinase type-2 alpha; phosphatidylinositol-4-phosphate 5-kinase, type II, alpha; phosphatidylinositol-5-phosphate 4-kinase type-2 alpha; phosphatidylinositol-5-phosphate 4-kinase, type II, alpha; PI(5)P 4-kinase type II alpha; PI5P4KA PIP5K2A PIP5KII-alpha PIP5KIIA PIPK 1-phosphatidylinositol-4-phosphate kinase 1-phosphatidylinositol-4-phosphate-5-kinase 1-phosphatidylinositol-5-phosphate 4-kinase 2-alpha PI(5)P 4-kinase type II alpha PIP4KII-alpha PIP5KIII PIP5KIIalpha PtdIns(4)P-5-kinase B isoform diphosphoinositide ki; PI5P4Kalpha; PIP4KII-alpha; PIP5KIIalpha; PIP5KIII; PIPK2 alpha; PtdIns(4)P-5-kinase B isoform; PtdIns(4)P-5-kinase C isoform; PtdIns(5)P-4-kinase isoform 2-alpha; type II phosphatidylinositol-4-phosphate 5-kinase 53 K isoform
Gene Aliases: AW742916; PI5P4KA; PIP4K2A; Pip4ka; PIP5K2; PIP5K2A; PIP5KII-alpha; PIP5KIIA; PIPK
UniProt ID: (Human) P48426, (Mouse) O70172, (Rat) Q9R0I8
Entrez Gene ID: (Human) 5305, (Mouse) 18718, (Rat) 116723
Molecular Function: kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.