Proteintech
Manufacturer Code:123681AP150UL
Catalog # 50-172-7931
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PINX1 Fusion Protein Ag3043 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 67-11-3 protein; hepatocellular carcinoma-related putative tumor suppressor; Liver-related putative tumor suppressor; LPTL LPTS PIN2 interacting protein 1 Pin2 interacting protein X1 PINX1 Protein 67 11 3 TRF1 interacting protein 1; LPTS1; PIN2 interacting protein 1; PIN2-interacting protein 1; PIN2-interacting protein X1; PIN2/TERF1-interacting telomerase inhibitor 1; PIN2/TRF1-interacting protein; pinx1 {ECO:0000312|EMBL:ABO28828.1}; Protein 67-11-3; TRF1-interacting protein 1
Gene Aliases: 2210403I16Rik; 2610028A01Rik; 67-11-3; AU024023; LPTL; LPTS; LPTS1; PINX1; RGD1566025
UniProt ID: (Human) Q96BK5, (Rat) A4L691, (Mouse) Q3V450
Entrez Gene ID: (Human) 54984, (Rat) 305963, (Mouse) 72400
Molecular Function:
enzyme modulator
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.