R&D Systems
Manufacturer Code:MAB4357SP
Catalog # MAB4357SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 420920 |
Immunogen | <EM>E. coli</EM>-derived recombinant human PINK1 Met1-Leu581 Accession # ABM87645 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: BRPK; BRPK EC 2.7.11.1 FLJ27236 PARK6 Parkinson disease (autosomal recessive) 6 protein kinase BRPK PTEN Induced Kinase 1 PTEN induced putative kinase 1 PTEN-induced putative kinase protein 1 serine/threonine-protein kinase PINK1 mitochondrial; protein kinase BRPK; PTEN-induced putative kinase protein 1; Serine/threonine-protein kinase PINK1, mitochondrial
Gene Aliases: BRPK; PARK6; PINK1
UniProt ID: (Human) Q9BXM7
Entrez Gene ID: (Human) 65018
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.