Invitrogen
Manufacturer Code:PA518770
Catalog # PIPA518770
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (QGKAHLESRSYQEAQ) corresponding to the internal amino acids of PINK1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: PINK1 PTEN induced putative kinase 1 BRPK FLJ27236 PARK6 protein kinase BRPK serine/threonine-protein kinase PINK1; serine/threonine-protein kinase PINK1, mitochondrial
Entrez Gene ID: (Rat) 298575
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.