Invitrogen
Manufacturer Code:PA513402
Catalog # PIPA513402
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 118-147 amino acids from the N-terminal region of human PINK1 (PARK6) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BRPK; protein kinase BRPK; PTEN induced putative kinase 1 protein kinase BRPK PINK1 Serine/threonine kinase PINK1 mitochondrial &lsqbPrecursor]; PTEN-induced putative kinase protein 1; Serine/threonine-protein kinase PINK1, mitochondrial
Gene Aliases: 1190006F07Rik; AU042772; AW557854; BRPK; mFLJ00387; PARK6; PINK1
UniProt ID: (Human) Q9BXM7, (Mouse) Q99MQ3
Entrez Gene ID: (Human) 65018, (Mouse) 68943
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.