Novus Biologicals
Manufacturer Code:NBP267796
Catalog # NBP267796
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | SD08-04 |
Immunogen | N terminal Human DYNLL1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
Protein Aliases: 8 kDa dynein light chain; cytoplasmic dynein light polypeptide; cytoplasmic dynein light polypeptide DLC1DNCLC1 DLC8MGC126137 DNCL1MGC126138 dynein light chain 1 cytoplasmic Dynein light chain LC8-type 1 dynein cytoplasmic light polypeptide 1 dynein light chain LC8-type 18 kDa dynein light chain hdlc1 LC8 PINLC8a Protein inhibitor of neuronal nitric oxide synthase; DLC8; Dynein light chain 1, cytoplasmic; Dynein light chain LC8-type 1; dynein, cytoplasmic, light polypeptide 1; dynein, light chain, LC8-type 1; PIN; Protein inhibitor of neuronal nitric oxide synthase
Gene Aliases: DLC1; DLC8; DNCL1; DNCLC1; DYNLL1; HDLC1; LC8; LC8a; PIN
UniProt ID: (Human) P63167
Entrez Gene ID: (Human) 8655
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.